Learn More
Cayman Chemical Fenspiride (hydrochloride), Purity: ≥95%, CAS: 5053-08-7, 5 g
SDP
Catalog No. 503015815
Encompass_Preferred
A histamine H1 receptor antagonist; inhibits histamine-induced contraction of isolated guinea pig trachea; inhibits PDE4, PDE5, and PDE3 (IC50s = 69, ~158, and 363 µM, respectively, in isolated human bronchi derived from patients with lung cancer); selective for these phosphodiesterases over PDE1 and PDE2; reverses bronchoconstriction induced by capsaicin at 1 mg/ml aerosolized and reduces cough in a guinea pig model of cough at 1-10 mg/ml
Catalog No. 50-301-5815 Supplier Cayman Chemical Supplier No. 255125 G
A histamine H1 receptor antagonist; inhibits histamine-induced contraction of isolated guinea pig trachea; inhibits PDE4, PDE5, and PDE3 (IC50s = 69, ~158, and 363 µM, respectively, in isolated human bronchi derived from patients with lung cancer); selective for these phosphodiesterases over PDE1 and PDE2; reverses bronchoconstriction induced by capsaicin at 1 mg/ml aerosolized and reduces cough in a guinea pig model of cough at 1-10 mg/ml