Learn More
Cliawaived Inc Fentanyl Rapid Tests
SDP
Supplier:  Cliawaived Inc AVRABCARD01FEN20
Encompass_Preferred
(25 Tests) Single Dipstick for Fentanyl (20 ng/mL cutoff) - FOR RESEARCH USE ONLY - Not FDA Cleared. Not CLIAwaived.
Catalog No. NC1859176