Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abbott™ Fentanyl Single Cassette RX

Catalog No. 22911198
Change view
Click to view available options
Quantity:
25 Cassettes/Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-911-198 25 Cassettes/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 22-911-198 Supplier Abbott™ Supplier No. DFYA102
This item is not returnable. View return policy

Fentanyl (FTY/FYL) 1 ng/mL urine cassette RX for drug screening. Moderately complex.

  • Compliant Fentanyl POCT solution
  • Adds relevant data for the clinician
  • Enhances drug test menu
  • Fast results leading to better diagnosis
  • Improved patient results due to fast POCT results

Specifications

CE Marker No
Clia Complexity High, Moderate
Detectable Analytes Fentanyl (FEN)
Format Cassette
Sample Type Urine
Sensitivity 1 ng/mL
Type Fentanyl Cassette
Disposable Single-use
For Use With (Application) Drug Screening
Quantity 25 Cassettes/Pk.
Form Ready-to-Use (RTU)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.