Learn More
BTNX Inc Rapid Response™ Fentanyl Test Cassette - 25 Tests
Catalog No. NC2945050
Encompass_Preferred
The Rapid Response™ Fentanyl Test Cassette detects the presence of fentanyl in human urine at the cut-off concentration of 1 ng/mL. This single drug test cassette can provide accurate and fast results in 5 minutes. The Rapid Response™ Fentanyl Test Cassette is CLIA Waived and is intended for use at the point-of-care. Kit contains 25 Tests per kit.The low cut-off level of 1 ng/mL enhances sensitivity for precise detection of fentanyl. This user-friendly fentanyl rapid test is designed for a seamless and effortless testing procedure proving results in just 5 minutes. This kit includes 25 individually packaged test cassettes sealed in a pouch, each includes a dropper for a mess-free and convenient testing procedure.
Catalog No. NC2945050 Supplier BTNX Inc Supplier No. FYL1CAP15625
The Rapid Response™ Fentanyl Test Cassette detects the presence of fentanyl in human urine at the cut-off concentration of 1 ng/mL. This single drug test cassette can provide accurate and fast results in 5 minutes. The Rapid Response™ Fentanyl Test Cassette is CLIA Waived and is intended for use at the point-of-care. Kit contains 25 Tests per kit.The low cut-off level of 1 ng/mL enhances sensitivity for precise detection of fentanyl. This user-friendly fentanyl rapid test is designed for a seamless and effortless testing procedure proving results in just 5 minutes. This kit includes 25 individually packaged test cassettes sealed in a pouch, each includes a dropper for a mess-free and convenient testing procedure.