Learn More
BTNX Inc Fentanyl Test Strip - 10pk (Liquid/Powder)
Catalog No. NC3361234
Encompass_Preferred
The Rapid Response™ Fentanyl Test Strip (Liquid/Powder) is an easy to use, cost-effective dipstick test. It is a rapid, qualitative screening test that quickly and reliably detects fentanyl at cut-off levels above 200 ng/mL. This drug test can provide accurate and fast results as soon as 1 minute. For forensic use only. Not an IVD.
Catalog No. NC3361234 Supplier BTNX Inc Supplier No. FYL18S710
The Rapid Response™ Fentanyl Test Strip (Liquid/Powder) is an easy to use, cost-effective dipstick test. It is a rapid, qualitative screening test that quickly and reliably detects fentanyl at cut-off levels above 200 ng/mL. This drug test can provide accurate and fast results as soon as 1 minute. For forensic use only. Not an IVD.