Learn More
BTNX Inc Rapid Response™ Fentanyl Test Strip (Liquid/Powder) - 100 Tests
Supplier:  BTNX Inc FYL18S7100
Encompass_Preferred
The Rapid Response™ Fentanyl Test Strip (Liquid/Powder) detects the presence of fentanyl in liquid and powder substances at a concentration of 200 ng/ml or higher. This drug test can provide accurate and fast results as soon as 1 minute. For forensic use only. Not an IVD. Kit contains 100 Tests per kit.WARNING: This test does not evaluate drug safety or purity.It is specific for fentanyl screening with no significant cross-reactivity to other opiates, such as morphine and heroin. This kit includes 100 individually packaged test strips sealed to prevent contamination, results interpretation card to aid in reading the results, and instructions. Every pouch features a QR code linking it to the BTNX Harm Reduction Resource Page – giving every user access to written and video instructions. The test results can be viewed in just 1 minute if the control line is present, however, it is recommended to wait 5 minutes after sample collection for best results.
Catalog No. NC2467570