Learn More
Chem Service Inc Fenthion | 55-38-9 | MFCD00055449 | 250 mg
SDP
Catalog No. NC1440286
Encompass_Preferred
This item has a minimum order quantity of 2 per supplier requirements
Fenthion | Purity: 99.5% | Mol Wt: 278.34 | 55-38-9 | MFCD00055449 | 250 mg
Catalog No. NC1440286 Supplier Chem Service Inc Supplier No. N11964250MG
This item has a minimum order quantity of 2 per supplier requirements
Fenthion | Purity: 99.5% | Mol Wt: 278.34 | 55-38-9 | MFCD00055449 | 250 mg