Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Fenthion-d6 | 1189662-83-6 | 95.0% | C10H9D6O3PS2 | 10MG
SDP

Catalog No. 5000260861
Encompass_Preferred

Fenthion-d6 is a deuterium-labeled analogue of the organophosphorus insecticide fenthion, provided as an analytical standard for tracer and quantitation in laboratory workflows. It is intended for research use only; handle and store according to the safety data sheet and the certificate of analysis.

  • Deuterium-labeled internal standard for accurate quantitation in mass spectrometry.
  • Suitable for LC-MS/MS and GC-MS method development and validation.
  • Molecular formula C10H9D6O3PS2 and molecular weight 284.37 g·mol⁻1 for clear identification.
  • Purity and storage conditions documented in the certificate of analysis.
  • Supplied in small research quantities appropriate for analytical use.

Catalog No. 50-002-60861 Supplier Medchemexpress LLC Supplier No. HY132457S10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Fenthion-d6 is a deuterium-labeled analogue of the organophosphorus insecticide fenthion, provided as an analytical standard for tracer and quantitation in laboratory workflows. It is intended for research use only; handle and store according to the safety data sheet and the certificate of analysis.

  • Deuterium-labeled internal standard for accurate quantitation in mass spectrometry.
  • Suitable for LC-MS/MS and GC-MS method development and validation.
  • Molecular formula C10H9D6O3PS2 and molecular weight 284.37 g·mol⁻1 for clear identification.
  • Purity and storage conditions documented in the certificate of analysis.
  • Supplied in small research quantities appropriate for analytical use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.