Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Fenthion-d6 | 1189662-83-6 | MFCD08273872 | >98.0% | 284.37 g/mol | C10H9D6O3PS2 | 5 MG
SDP

Catalog No. 5000263225
Encompass_Preferred

Fenthion-d6 is a deuterium-labeled analogue of the organophosphorus insecticide fenthion provided as a research standard for analytical and tracer applications. It is intended for laboratory use in method development, calibration, and quantitative mass spectrometry workflows; purity and recommended storage conditions are referenced in the product Certificate of Analysis.

  • Deuterium-labeled tracer for LC-MS/MS and GC-MS applications.
  • Enables accurate quantitation when used as an internal standard.
  • Molecular formula C10H9D6O3PS2; molecular weight 284.37 g/mol.
  • Available in small milligram pack sizes suitable for analytical standards.
  • For research use only; refer to the Certificate of Analysis for purity and storage.

Catalog No. 50-002-63225 Supplier Medchemexpress LLC Supplier No. HY132457S5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Fenthion-d6 is a deuterium-labeled analogue of the organophosphorus insecticide fenthion provided as a research standard for analytical and tracer applications. It is intended for laboratory use in method development, calibration, and quantitative mass spectrometry workflows; purity and recommended storage conditions are referenced in the product Certificate of Analysis.

  • Deuterium-labeled tracer for LC-MS/MS and GC-MS applications.
  • Enables accurate quantitation when used as an internal standard.
  • Molecular formula C10H9D6O3PS2; molecular weight 284.37 g/mol.
  • Available in small milligram pack sizes suitable for analytical standards.
  • For research use only; refer to the Certificate of Analysis for purity and storage.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.