Learn More
Medchemexpress LLC Fenticonazole Nitrate | 73151-29-8 | 98.86% | 100 MG
SDP
Catalog No. 5000407307
Encompass_Preferred
Fenticonazole Nitrate is an antifungal imidazole ring derivative that primarily functions by hindering ergosterol integration, leading to the destruction of the cytoplasmic outer membrane. This action makes it effective against a range of pathogens.
  • Effective against Gram-positive bacteria, mycoses, and vaginal candidiasis
  • Active against dermatophyte pathogens, Malassezia furfur, and Candida albicans
  • Used locally in the treatment of vulvovaginal candidiasis
  • For research use only
Catalog No. 50-004-07307 Supplier Medchemexpress LLC Supplier No. HYB0359100MG
Fenticonazole Nitrate is an antifungal imidazole ring derivative that primarily functions by hindering ergosterol integration, leading to the destruction of the cytoplasmic outer membrane. This action makes it effective against a range of pathogens.
  • Effective against Gram-positive bacteria, mycoses, and vaginal candidiasis
  • Active against dermatophyte pathogens, Malassezia furfur, and Candida albicans
  • Used locally in the treatment of vulvovaginal candidiasis
  • For research use only