Learn More
Medchemexpress LLC Fenticonazole Nitrate | 73151-29-8 | 98.86% | 50 MG
SDP
Catalog No. 5000406608
Encompass_Preferred
Fenticonazole Nitrate is an antifungal imidazole ring derivative that hinders ergosterol integration and destructs the cytoplasmatic outer membrane. It is effective against Gram-positive bacteria, mycoses, and vaginal candidiasis.
  • Antifungal imidazole ring derivative.
  • Operates by hindering ergosterol integration.
  • Destructs the cytoplasmatic outer membrane.
  • Effective against Gram-positive bacteria.
  • Effective against mycoses.
  • Effective against vaginal candidiasis.
Catalog No. 50-004-06608 Supplier Medchemexpress LLC Supplier No. HYB035950MG
Fenticonazole Nitrate is an antifungal imidazole ring derivative that hinders ergosterol integration and destructs the cytoplasmatic outer membrane. It is effective against Gram-positive bacteria, mycoses, and vaginal candidiasis.
  • Antifungal imidazole ring derivative.
  • Operates by hindering ergosterol integration.
  • Destructs the cytoplasmatic outer membrane.
  • Effective against Gram-positive bacteria.
  • Effective against mycoses.
  • Effective against vaginal candidiasis.