Learn More
Cayman Chemical FentIconazolenItrate 10mg
SDP
Catalog No. 502938100
Encompass_Preferred
An antimicrobial imidazole; active against clinical isolates of Candida species (MICs = 0.12-32 μg/ml); active against clinical isolates of bacteria associated with bacterial vaginosis (MICs = ≤0.03-16 μg/ml), as well as bacteria associated with skin infections (MICs = ≤0.03->16 μg/ml); inhibits fungal growth in a guinea pig model of experimental M. canis dermatomycosis (ED50 = 0.5%, applied topically)
Catalog No. 50-293-8100 Supplier Cayman Chemical Supplier No. 2900210 MG
An antimicrobial imidazole; active against clinical isolates of Candida species (MICs = 0.12-32 μg/ml); active against clinical isolates of bacteria associated with bacterial vaginosis (MICs = ≤0.03-16 μg/ml), as well as bacteria associated with skin infections (MICs = ≤0.03->16 μg/ml); inhibits fungal growth in a guinea pig model of experimental M. canis dermatomycosis (ED50 = 0.5%, applied topically)