Learn More
Cayman Chemical Fenticonazole (nitrate), Purity: ≥98%, CAS: 73151-29-8, 50 mg
SDP
Catalog No. 502940948
Encompass_Preferred
An antimicrobial imidazole; active against clinical isolates of Candida species (MICs = 0.12-32 μg/ml); active against clinical isolates of bacteria associated with bacterial vaginosis (MICs = ≤0.03-16 μg/ml), as well as bacteria associated with skin infections (MICs = ≤0.03->16 μg/ml); inhibits fungal growth in a guinea pig model of experimental M. canis dermatomycosis (ED50 = 0.5%, applied topically)
Catalog No. 50-294-0948 Supplier Cayman Chemical Supplier No. 2900250 MG
An antimicrobial imidazole; active against clinical isolates of Candida species (MICs = 0.12-32 μg/ml); active against clinical isolates of bacteria associated with bacterial vaginosis (MICs = ≤0.03-16 μg/ml), as well as bacteria associated with skin infections (MICs = ≤0.03->16 μg/ml); inhibits fungal growth in a guinea pig model of experimental M. canis dermatomycosis (ED50 = 0.5%, applied topically)