Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Fentonium bromide | 5868-06-4 | 98.8% | 10 MG
SDP

Catalog No. 5000230970
Encompass_Preferred

Fentonium bromide is an anticholinergic, antispasmodic, and anti-ulcerogenic research chemical used in studies of neurological conditions such as unstable bladder. It is supplied for laboratory research in small solid quantities.

  • Purity 98.8%.
  • Molecular weight 564.51 g/mol.
  • Chemical formula C31H34BrNO4.
  • Available in milligram-scale solid quantities for research use.
  • Used in research on anticholinergic activity and bladder instability.

Catalog No. 50-002-30970 Supplier Medchemexpress LLC Supplier No. HY12252010MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Fentonium bromide is an anticholinergic, antispasmodic, and anti-ulcerogenic research chemical used in studies of neurological conditions such as unstable bladder. It is supplied for laboratory research in small solid quantities.

  • Purity 98.8%.
  • Molecular weight 564.51 g/mol.
  • Chemical formula C31H34BrNO4.
  • Available in milligram-scale solid quantities for research use.
  • Used in research on anticholinergic activity and bladder instability.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.