Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Fentonium bromide | 5868-06-4 | 98.8% | 10MM 1ML
SDP

Catalog No. 5000230245
Encompass_Preferred

Fentonium bromide | 5868-06-4 | 98.8% | 10MM 1ML

Catalog No. 50-002-30245 Supplier Medchemexpress LLC Supplier No. HY12252010MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Fentonium bromide | 5868-06-4 | 98.8% | 10MM 1ML

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.