Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Fentonium bromide | 5868-06-4 | 98.8% | 5 MG
SDP

Catalog No. 5000233721
Encompass_Preferred

Fentonium bromide is an anticholinergic and antispasmodic research compound used in pharmacological studies of gastrointestinal and neurological conditions, including unstable bladder. Supplied as the bromide salt in solid form, it is commonly used as a tool compound in studies of muscarinic acetylcholine receptors and is typically dissolved in DMSO for experimental use.

  • CAS 5868-06-4
  • Chemical formula C31H34BrNO4
  • Molecular weight 564.51 g/mol
  • Purity 98.75%
  • Form bromide salt, solid
  • Solubility soluble in DMSO
  • Intended use research reagent targeting muscarinic acetylcholine receptors
  • Available in small quantities such as 5 mg

Catalog No. 50-002-33721 Supplier Medchemexpress LLC Supplier No. HY1225205MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Fentonium bromide is an anticholinergic and antispasmodic research compound used in pharmacological studies of gastrointestinal and neurological conditions, including unstable bladder. Supplied as the bromide salt in solid form, it is commonly used as a tool compound in studies of muscarinic acetylcholine receptors and is typically dissolved in DMSO for experimental use.

  • CAS 5868-06-4
  • Chemical formula C31H34BrNO4
  • Molecular weight 564.51 g/mol
  • Purity 98.75%
  • Form bromide salt, solid
  • Solubility soluble in DMSO
  • Intended use research reagent targeting muscarinic acetylcholine receptors
  • Available in small quantities such as 5 mg

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.