Learn More
Cayman Chemical Fenvalerate, Purity: ≥95%, CAS: 51630-58-1, 100 mg
SDP
Catalog No. 502989371
Encompass_Preferred
A pyrethroid insecticide and acaricide; a slow activator of Nav1.8; induces mortality of pyrethroid-susceptible and -resistant strains in M. domestica (LD50s = 0.014-5 µg/fly); induces mortality in tobacco budworm larvae but is associated with pyrethroid resistance with an increase in LD50 values from 1.01 to 20.85 µg/g over a six-year timeframe; induces mortality in 95.5% of T. macfarlanei spider mites at 0.015%
Catalog No. 50-298-9371 Supplier Cayman Chemical Supplier No. 29399100 MG
A pyrethroid insecticide and acaricide; a slow activator of Nav1.8; induces mortality of pyrethroid-susceptible and -resistant strains in M. domestica (LD50s = 0.014-5 µg/fly); induces mortality in tobacco budworm larvae but is associated with pyrethroid resistance with an increase in LD50 values from 1.01 to 20.85 µg/g over a six-year timeframe; induces mortality in 95.5% of T. macfarlanei spider mites at 0.015%