Learn More
Chem Service Inc Fenvalerate, 51630-58-1, MFCD00055324, 250 mg
SDP
Catalog No. NC1569792
Encompass_Preferred
Molecular Formula: C25-H22-Cl-N-O3, Molecular Weight: 419.92, Classification: Insecticide, Acaricide, Ixodicide, Alternate Name(s): Sanmarton.
Catalog No. NC1569792 Supplier Chem Service Inc Supplier No. N13201250MG
Molecular Formula: C25-H22-Cl-N-O3, Molecular Weight: 419.92, Classification: Insecticide, Acaricide, Ixodicide, Alternate Name(s): Sanmarton.