Learn More
Cole-Parmer Digi-Sense Type-K, FEP-Insulated Probe, Coated Junct, Mini-Connector, Ungrounded, 10ft 24 -Gauge
Catalog No. NC1614137 Shop All Cole-Parmer Products
Encompass_Preferred
Insulated with FEP-sealed junction this probe is used for long-term immersion in semi-caustic environments. Each probe is hermetically sealed and tested to 25 psi. Can be used for applications requiring an isolated junction. Response Time: 0.75 seconds or greater. All insulated-wire probes include straight insulated cable with no handle and feature ANSI color-coded mini connectors: type J - black, type K - yellow, and type T - blue.
Catalog No. NC1614137 Supplier Cole-Parmer Supplier No. 0846682
Insulated with FEP-sealed junction this probe is used for long-term immersion in semi-caustic environments. Each probe is hermetically sealed and tested to 25 psi. Can be used for applications requiring an isolated junction. Response Time: 0.75 seconds or greater. All insulated-wire probes include straight insulated cable with no handle and feature ANSI color-coded mini connectors: type J - black, type K - yellow, and type T - blue.