Learn More
Analytical Sales & Services FEP Micro Tubing Sleeve for .039" (1mm) OD Tubing
Catalog No. 5000804713
Encompass
Silica Seal Tight FEP Tubing Sleeve\n\n• Length: 1.6"\n• Inner Diameter: .042", for 0.0394" (1mm) OD Tubing\n• Outer Diameter: 1/16"\n• Color: Purple
Catalog No. 50-008-04713 Supplier Analytical Sales & Services Supplier No. 55252
Silica Seal Tight FEP Tubing Sleeve\n\n• Length: 1.6"\n• Inner Diameter: .042", for 0.0394" (1mm) OD Tubing\n• Outer Diameter: 1/16"\n• Color: Purple