Learn More
Analytical Sales & Services FEP Micro Tubing Sleeve for 785-825μm OD Tubing
Catalog No. 5000804712
Encompass
Silica Seal Tight FEP Micro Tubing Sleeve\n\n• Length: 1.6"\n• Inner Diameter: 840μm (0.033"), for 785-825 μm OD Tubing\n• Outer Diameter: 1/16"\n• Color: Green
Catalog No. 50-008-04712 Supplier Analytical Sales & Services Supplier No. 55247
Silica Seal Tight FEP Micro Tubing Sleeve\n\n• Length: 1.6"\n• Inner Diameter: 840μm (0.033"), for 785-825 μm OD Tubing\n• Outer Diameter: 1/16"\n• Color: Green