Learn More
Analytical Sales & Services FEP Teflon Tubing, 1/16" OD x 0.020" ID x 50'
Catalog No. 5000804634
Encompass
FEP Teflon Tubing\n\nOuter Diameter: 1/16”\nInner Diameter: 0.020”\nLength: 50’\n\nQty/Pkg: Each
Catalog No. 50-008-04634 Supplier Analytical Sales & Services Supplier No. 41548L4
FEP Teflon Tubing\n\nOuter Diameter: 1/16”\nInner Diameter: 0.020”\nLength: 50’\n\nQty/Pkg: Each