Learn More
Analytical Sales & Services FEP Tubing, 6mm OD x 4mm ID x 1mm Wall Thickness
Catalog No. 5000804697
Encompass
FEP Teflon Tubing\n\n\n\n• Outer Diameter: 6mm\n\n• Inner Diameter: 4mm\n\n• Length: 100'\n\n\n\nQty/Pkg: Each
Catalog No. 50-008-04697 Supplier Analytical Sales & Services Supplier No. 4138100
FEP Teflon Tubing\n\n\n\n• Outer Diameter: 6mm\n\n• Inner Diameter: 4mm\n\n• Length: 100'\n\n\n\nQty/Pkg: Each