Learn More
Ace Glass, Inc. FEP tubing, 7/16in OD x 3/8in ID, 10ft length
SDP
Catalog No. 50983384
Encompass
7/16 O.D x 3/8 I.D. FEP tubing made of high molecular weight resin for excellent stress crack resistance. Meets FDA requirements for repeated food contact. Chemically inert to most industrial chemicals and solvents. 10ft length.
Catalog No. 50-983-384 Supplier Ace Glass, Inc. Supplier No. 1268720
7/16 O.D x 3/8 I.D. FEP tubing made of high molecular weight resin for excellent stress crack resistance. Meets FDA requirements for repeated food contact. Chemically inert to most industrial chemicals and solvents. 10ft length.