Learn More
Perkin Elmer US LLC FEP Tubing - Long Screw Fitting, 500 mm
Catalog No. 502104401
Encompass_Preferred
This FEP Tubing is a flexible, heat-resistant, transparent tube that is 500 mm long. It has one long screw fitting. It is a replacement tube for UV/Vis Flow-Through cells. With this part number, you can order this tube as a spare. Qty. 1
Catalog No. 50-210-4401 Supplier Perkin Elmer US LLC Supplier No. B0631131
This FEP Tubing is a flexible, heat-resistant, transparent tube that is 500 mm long. It has one long screw fitting. It is a replacement tube for UV/Vis Flow-Through cells. With this part number, you can order this tube as a spare. Qty. 1