Learn More
Medchemexpress LLC Feprazone | 25MG
SDP
Catalog No. 5000307871
Encompass_Preferred
Feprazone (DA2370 Prenazone) an analogue of Phenylbutazone (HY-B0230) is a nonsteroidal anti-inflammatory agent with analgesic and antipyretic activities Feprazone acts by inhibiting the activity of cyclooxygenase (COX)-2 Feprazone ameliorates free fatty acid (FFA)-induced oxidative stress by reducing the production of mitochondrial reactive oxygen species (ROS) Feprazone can decrease the expression of MMP-2 and MMP-9 Besides Feprazone can suppress adipogenesis and increase lipolysis in differentiating 3 T3-L1 cells Feprazone also can be used to research atherosclerosis and obesity[1][2][3]
Catalog No. 50-003-07871 Supplier Medchemexpress LLC Supplier No. HY11491125MG
Feprazone (DA2370 Prenazone) an analogue of Phenylbutazone (HY-B0230) is a nonsteroidal anti-inflammatory agent with analgesic and antipyretic activities Feprazone acts by inhibiting the activity of cyclooxygenase (COX)-2 Feprazone ameliorates free fatty acid (FFA)-induced oxidative stress by reducing the production of mitochondrial reactive oxygen species (ROS) Feprazone can decrease the expression of MMP-2 and MMP-9 Besides Feprazone can suppress adipogenesis and increase lipolysis in differentiating 3 T3-L1 cells Feprazone also can be used to research atherosclerosis and obesity[1][2][3]