Learn More
Medchemexpress LLC Feprazone 5mg | 30748-29-9 | 5 MG
SDP
Catalog No. 5000227708
Encompass_Preferred
Feprazone is a pyrazolone nonsteroidal anti-inflammatory agent provided as an analytical standard for research and analytical applications. It exhibits analgesic and antipyretic activity and is used in studies of COX-2 inhibition, oxidative stress, and adipogenesis.
  • Analytical standard for research and quality control
  • Used to study COX-2 inhibition and oxidative stress
  • Molecular formula C20H20N2O2 and molecular weight 320.4 g/mol
  • Suitable for biochemical and pharmacological assays
Catalog No. 50-002-27708 Supplier Medchemexpress LLC Supplier No. HY1149115MG
Feprazone is a pyrazolone nonsteroidal anti-inflammatory agent provided as an analytical standard for research and analytical applications. It exhibits analgesic and antipyretic activity and is used in studies of COX-2 inhibition, oxidative stress, and adipogenesis.
  • Analytical standard for research and quality control
  • Used to study COX-2 inhibition and oxidative stress
  • Molecular formula C20H20N2O2 and molecular weight 320.4 g/mol
  • Suitable for biochemical and pharmacological assays