Learn More
Neta Scientific Agilent Ferrule, 0.4mm G 0.05-0.25 col 10/PK
SDP
Catalog No. 50926934
Encompass_Preferred
This item is manufactured by Agilent
Agilent Ferrule, 0.4 mm id, graphite, 0.05 to 0.25 mm column, 10/pk - Graphite Ferrules for GC
UNSPSC Code  41115717
- 58
Catalog No. 50-926-934 Supplier Neta Scientific Supplier No. 5002114
This item is manufactured by Agilent
Agilent Ferrule, 0.4 mm id, graphite, 0.05 to 0.25 mm column, 10/pk - Graphite Ferrules for GC
UNSPSC Code  41115717
- 58