Learn More
Neta Scientific Agilent Ferrule, 0.5mm Graphite 0.32 col 10/PK
SDP
Catalog No. 50926879
Encompass_Preferred
This item is manufactured by Agilent
Agilent Ferrule, graphite, short, 0.5 mm id, for 0.1 to 0.32 mm columns, 10/pk - Cool On-Column (COC) Inlet for GC
UNSPSC Code  41115717
- 58
This item is not returnable. View return policy
Catalog No. 50-926-879 Supplier Neta Scientific Supplier No. 50808853
This item is manufactured by Agilent
Agilent Ferrule, graphite, short, 0.5 mm id, for 0.1 to 0.32 mm columns, 10/pk - Cool On-Column (COC) Inlet for GC
UNSPSC Code  41115717
- 58