Learn More
Neta Scientific Agilent Ferrule, 1.0mm Graphite 0.53 col 10/PK
SDP
Catalog No. 50926946
Encompass_Preferred
This item is manufactured by Agilent
Agilent Ferrule, graphite, 1 mm id, for 0.53 mm capillary columns, 10/pk - Graphite Ferrules
UNSPSC Code  41115717
- 58
Catalog No. 50-926-946 Supplier Neta Scientific Supplier No. 50808773
This item is manufactured by Agilent
Agilent Ferrule, graphite, 1 mm id, for 0.53 mm capillary columns, 10/pk - Graphite Ferrules
UNSPSC Code  41115717
- 58