Learn More
Trajan Scientific FER GVS-I-004 QP5000 MSI 10/PK
SDP
Catalog No. 502550761
Encompass_Preferred
SGE 15% Graphite/85% Vespel Ferrules, Diameter Inside: 0.010 to 0.0098 in., 0.0039 to 0.025 cm, For Use With: GC, QP5000/5050 Standard MS Interface, 15% Graphite, 85% Vespel
Catalog No. 50-255-0761 Supplier Trajan Scientific Supplier No. 0726563
SGE 15% Graphite/85% Vespel Ferrules, Diameter Inside: 0.010 to 0.0098 in., 0.0039 to 0.025 cm, For Use With: GC, QP5000/5050 Standard MS Interface, 15% Graphite, 85% Vespel