Learn More
Biocare Medical, LLC Ferangi Blue Chromogen Kit 2 - 25 ml
SDP
Catalog No. 5082836
Encompass_Preferred
The Ferangi Blue Chromogen Kit 2 is provided as a stable, two-component system, consisting of Ferangi Blue Chromogen and Ferangi Blue Buffer. When in the presence of alkaline phosphatase (AP) enzyme, Ferangi Blue Chromogen Kit 2 produces a permanent, bright royal blue precipitate. Typically, Ferangi Blue achieves robust staining intensity in 5-7 minutes. If more intense staining is desired, Ferangi Blue can be extended to 10 minutes, offering users increased flexibility and sensitivity. Ferangi Blue is particularly well-suited for automated multiplex staining applications. Ferangi Blue is partially soluble in xylenes. Slides should be air dried and coverslipped in a xylene-substitute permanent mounting medium, such as Biocare EcoMount (EM897).
Catalog No. 50-828-36 Supplier Biocare Medical, LLC Supplier No. FB813H
The Ferangi Blue Chromogen Kit 2 is provided as a stable, two-component system, consisting of Ferangi Blue Chromogen and Ferangi Blue Buffer. When in the presence of alkaline phosphatase (AP) enzyme, Ferangi Blue Chromogen Kit 2 produces a permanent, bright royal blue precipitate. Typically, Ferangi Blue achieves robust staining intensity in 5-7 minutes. If more intense staining is desired, Ferangi Blue can be extended to 10 minutes, offering users increased flexibility and sensitivity. Ferangi Blue is particularly well-suited for automated multiplex staining applications. Ferangi Blue is partially soluble in xylenes. Slides should be air dried and coverslipped in a xylene-substitute permanent mounting medium, such as Biocare EcoMount (EM897).