Learn More
Aviva Systems Biology Corp FERD3L Antibody - N-terminal region (ARP36206_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP36206_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against FERD3L. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human FERD3L. Alternate names: NATO3, NTWIST, N-TWIST, bHLHa31ApplicationWB. Species reactivity: Human
Catalog No. 50-014-56065