Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ferene disodium | 79551-14-7 | 99.94% | 250 MG
SDP

Catalog No. 5000414242
Encompass_Preferred

Ferene disodium is a biochemical reagent utilized in life science research as a biological material or organic compound. It is intended for research use only and is not for sale to patients. This compound has a molecular weight of 494.37 and a chemical formula of C16H8N4Na2O8S2. It presents as a light yellow to yellow solid and can be stored under various conditions depending on its state.

  • Used as a biochemical reagent for life science research
  • Intended for research use only
  • Molecular weight of 494.37
  • Chemical formula is C16H8N4Na2O8S2
  • Appears as a light yellow to yellow solid
  • Store at room temperature in continental US or 4°C sealed, away from moisture
  • In solvent, store at -80°C for 6 months or -20°C for 1 month
  • Soluble in H2O (100 mg/mL) and DMSO (100 mg/mL)

Catalog No. 50-004-14242 Supplier Medchemexpress LLC Supplier No. 250MGHYD0170
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ferene disodium is a biochemical reagent utilized in life science research as a biological material or organic compound. It is intended for research use only and is not for sale to patients. This compound has a molecular weight of 494.37 and a chemical formula of C16H8N4Na2O8S2. It presents as a light yellow to yellow solid and can be stored under various conditions depending on its state.

  • Used as a biochemical reagent for life science research
  • Intended for research use only
  • Molecular weight of 494.37
  • Chemical formula is C16H8N4Na2O8S2
  • Appears as a light yellow to yellow solid
  • Store at room temperature in continental US or 4°C sealed, away from moisture
  • In solvent, store at -80°C for 6 months or -20°C for 1 month
  • Soluble in H2O (100 mg/mL) and DMSO (100 mg/mL)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.