Learn More
Broadley James Corporation F-695 Autoclavable pH Electrode, use with/3L-5L Vessel, 225 mm
Catalog No. NC9926376
Encompass
Model F-695 pH FermProbe with Standard Metric CapThe F-695 is similar to the F-635 and varies only in the connector type. Suitable for either benchtop or production vessels. Steam sterilizable and autoclavable.Use with/3L-5L Vessel, 225 mm 0- 13 pH rangeSteam sterilizable to 135° C150 psig max. pressureDouble junction, Ag-AgCl reference systemMetric Pg13.5 threaded disconnect capNote: the vessel sizes are for guidance only – check your own vessel dimensions or the length of the electrode you are replacing.
Catalog No. NC9926376 Supplier Broadley James Corporation Supplier No. F695B225DK
Model F-695 pH FermProbe with Standard Metric CapThe F-695 is similar to the F-635 and varies only in the connector type. Suitable for either benchtop or production vessels. Steam sterilizable and autoclavable.Use with/3L-5L Vessel, 225 mm 0- 13 pH rangeSteam sterilizable to 135° C150 psig max. pressureDouble junction, Ag-AgCl reference systemMetric Pg13.5 threaded disconnect capNote: the vessel sizes are for guidance only – check your own vessel dimensions or the length of the electrode you are replacing.