Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ DecaLabel DNA Labeling Kit

Catalog No. FERK0622 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
30 Reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FERK0622 30 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FERK0622 Supplier Thermo Scientific™ Supplier No. K0622

missing translation for 'validMainframeMsg'

Thermo Scientific™ Fermentas DecaLabel DNA Labeling Kit is a complete system for fast synthesis of radiolabeled DNA probes of high specific activity.

Perform fast synthesis of radiolabeled DNA probes of high specific activity with the complete system provided by Thermo Scientific™ Fermentas DecaLabel™ DNA Labeling Kit.

The kit is based on the improved random-primed method developed by Feinberg and Vogelstein (1, 2). The primary improvement over traditional random-primer kits involves the use of random decamers instead of hexamers to ensure more efficient annealing with DNA at 37°C. In addition, Klenow Fragment, exo- has been genetically engineered to remove exonuclease activity. The improved enzyme does not degrade the labeled probe during reaction and allows high labeling yields even from low template amounts. DNA fragments of any length can be uniformly labeled. Two labeling mixes are provided for flexibility in using either radiolabeled dATP or dCTP.

• High specific radioactivity of probes—1 x 109 dpm/μg DNA
• Fast—5 minutes at 37°C
• High yields with Klenow Fragment, exo-: no degradation of a labeled probe during reaction
• Flexible—suitable for either radiolabeled dATP or dCTP

Recommended for:
Generation of radiolabeled DNA probes for use in a variety of hybridization experiments: Southern and Northern blots, colony/plaque hybridizations, dot/slot blots and in situ hybridizations.

TRUSTED_SUSTAINABILITY

Specifications

Detection Method Radioactive
Format Kit
Final Product Type DNA
Labeling Target Genomic DNA, PCR Products, DNA (General)
Includes Label or Dye No
Labeling Method Direct Labeling
Product Line DecaLabel
Product Type DNA Labeling Kit
Quantity 30 Reactions

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.