Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ pTZ19R DNA

Catalog No. FERSD0141
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FERSD0141 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FERSD0141 Supplier Thermo Scientific™ Supplier No. SD0141
Add to Cart
Add to Cart

Perform DNA cloning, dideoxy DNA sequencing, in vitro mutagenesis, and in vitro transcription in one system with this double-stranded closed circular high copy phagemid DNA.

pTZ19R/U are small phagemids 2862 bp in length, constructed by inserting the DNA of phage f1 intergenic region (IG) into pUC19 and the T7 promoter sequence near to the MCS of pUC19. The phagemids differ in the orientation of the cloned f1 IG region. They are designed for DNA cloning, dideoxy DNA sequencing, in vitro mutagenesis, and in vitro transcription in one system. A host strain harboring these phagemids will produce single-stranded DNA if superinfected with the helper phage M13K07.

Highlights

  • Isolated from E. coli (dam+, dcm+)
  • More than 90% in the supercoiled form
  • Isolated from E.coli (dam+, dcm-)
TRUSTED_SUSTAINABILITY

Specifications

Cloning Method Restriction Enzyme⁄ MCS
Concentration 0.5 μg/μL
Sample Type DNA
Final Product Type DNA
For Use With (Application) Cloning
Product Type DNA
Quantity 50 μg
Vector pTZ19R
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.