Learn More
Thermo Scientific™ pUC19 DNA/MspI (HpaII) Marker, ready-to-use
GREENER_CHOICE
Catalog No. FERSM0223 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
50 μg
Catalog No. Quantity
FERSM0223 50 μg
1 options
Catalog No. FERSM0223 Supplier Thermo Scientific™ Supplier No. SM0223
Add to Cart
Add to Cart
Size and approximately quantify small DNA fragments in agarose and non-denaturing polyacrylamide gels. Premixed with Loading Dye for direct loading onto gel.
0.5 μg/lane, 8 cm length gel, 1X TBE, 5 V/cm, 1.5 h. Bands visible: 501, 489, 404, 331, 242, 190, 147, 111, 110, 67, 34 and 34 bp.
pUC19 DNA is digested to completion with the appropriate Thermo Scientific restriction enzyme, then purified and dissolved in storage buffer. The DNA fragments contain blunt or sticky ends, depending on the restriction enzyme used for the markers preparation. pUC19 DNA/MspI (HpaII) Marker, ready-to-use, is premixed with 6X DNA Loading Dye for direct loading onto agarose and polyacrylamide gels.
Highlights
  • Sizing and approximate quantification of small DNA fragments
  • Sharp bands 
  • Premixed with 6X DNA Loading Dye for direct loading and room temperature storage
  • Supplied with loading dye for sample DNA
TRUSTED_SUSTAINABILITY
Specifications
Concentration 6X
Gel Type Agarose
Ready to Load Yes
Gel Compatibility Agarose Gels
Quantity 50 μg
Product Type DNA/MSPI (HPAII) Marker
What is the recommended voltage for electrophoresis with Thermo Scientific DNA and RNA ladders? What does V/cm mean?
Each of Thermo Scientific DNA and RNA ladders has a recommended voltage given in V/cm. Please refer to the gel picture in the product information sheet. V/cm stands for the amount of voltage recommended per cm of the distance between the two electrodes of the gel box (not gel length).
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
For Research Use Only. Not for use in diagnostic procedures.