Learn More
Bellco Glass, Inc. Fermentation Tube, 8x29 mm
SDP
Catalog No. 501945334
Encompass_Preferred
Disposable gas fermentation tubes are inverted in bottom of larger tube for collection of fermentation gases. Disposible standard flint glass with 1.15mm Wall. 144 per case. Size: 8x29mm
Catalog No. 50-194-5334 Supplier Bellco Glass, Inc. Supplier No. 204008029
Disposable gas fermentation tubes are inverted in bottom of larger tube for collection of fermentation gases. Disposible standard flint glass with 1.15mm Wall. 144 per case. Size: 8x29mm