Learn More
Broadley James Corporation F-235 Sterilizable pH Electrode, pH Range: 0-13 pH, Temperature Range: 0-135 ˚C
Catalog No. NC0219494
Encompass
The F-235 Sterilizable pH Electrode is a sealed, gel-filled, sterilizable, autoclavable design with built-in temperature element. This allows for the automatic compensation of Nernstian temperature effects in processes with wide pH ranges and significant temperature shifts.The VarioPin connector allows for a multiple connection; both pH signal and temperature signal in the same wiring harness. The temperature element is either a PT100 or PT1000 Platinum Resistance Device, which are compatible with the majority of commercially available instruments. The unique construction methods used by Broadley-James allows this sterilizable pH electrode to operate at pressures of up to 10bar (150psig) and temperatures up to 135°C.F-235 Sterilizable pH Electrode SpecificationspH Range: 0-13 pHTemperature Range: 0-135 ˚CTemperature Range: 32-275 ˚FPressure Range: 0-150 psigPressure Range: 0-10 barTemperature Sensor Range: 0-100 ˚C
Catalog No. NC0219494 Supplier Broadley James Corporation Supplier No. F235D120VP
The F-235 Sterilizable pH Electrode is a sealed, gel-filled, sterilizable, autoclavable design with built-in temperature element. This allows for the automatic compensation of Nernstian temperature effects in processes with wide pH ranges and significant temperature shifts.The VarioPin connector allows for a multiple connection; both pH signal and temperature signal in the same wiring harness. The temperature element is either a PT100 or PT1000 Platinum Resistance Device, which are compatible with the majority of commercially available instruments. The unique construction methods used by Broadley-James allows this sterilizable pH electrode to operate at pressures of up to 10bar (150psig) and temperatures up to 135°C.F-235 Sterilizable pH Electrode SpecificationspH Range: 0-13 pHTemperature Range: 0-135 ˚CTemperature Range: 32-275 ˚FPressure Range: 0-150 psigPressure Range: 0-10 barTemperature Sensor Range: 0-100 ˚C