Learn More
Broadley James Corporation F-635 Autoclavable pH Electrode, Use with/ 3L-5L Vessel 225 mm
Catalog No. NC9966548
Encompass
This FermProbe autoclavable pH electrode style features a metric Pg13.5 threaded polymer cap that allows it to be used with European style electrode housings. Autoclavable as well as steam sterilizable, this style of pH FermProbe is the first choice for use with small autoclavable benchtop vessels. Available only in the disconnect cap style. Cables must be ordered separately (see the Accessories tab for compatible cables).Features:Metric threaded cap is compatible with nearly all European designed bioprocess electrode housings.Steam sterilizable and autoclavable.First choice for use with small autoclavable vessels.Available in variety of lengths for different insertion length requirements and applications.0 – 13 pH rangeSteam sterilizable to 135° C150 psig max. pressureDouble junction, Ag-AgCl reference systemMetric Pg13.5 threaded disconnect capNote: the vessel sizes are for guidance only – check your own vessel dimensions or the length of the electrode you are replacing.
Catalog No. NC9966548 Supplier Broadley James Corporation Supplier No. F635B225DH
This FermProbe autoclavable pH electrode style features a metric Pg13.5 threaded polymer cap that allows it to be used with European style electrode housings. Autoclavable as well as steam sterilizable, this style of pH FermProbe is the first choice for use with small autoclavable benchtop vessels. Available only in the disconnect cap style. Cables must be ordered separately (see the Accessories tab for compatible cables).Features:Metric threaded cap is compatible with nearly all European designed bioprocess electrode housings.Steam sterilizable and autoclavable.First choice for use with small autoclavable vessels.Available in variety of lengths for different insertion length requirements and applications.0 – 13 pH rangeSteam sterilizable to 135° C150 psig max. pressureDouble junction, Ag-AgCl reference systemMetric Pg13.5 threaded disconnect capNote: the vessel sizes are for guidance only – check your own vessel dimensions or the length of the electrode you are replacing.