Learn More
Aviva Systems Biology Corp FERMT1 Antibody - N-terminal region (ARP42485_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP42485_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against FERMT1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human FERMT1. Alternate names: C20orf42, DTGCU2, FLJ20116, FLJ23423, KIND1, URP1, unc112a, UNC112AApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Rabbit, Rat
Catalog No. 50-014-53338