Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ferno™ EZ Glide™ Evacuation Chair: Accessories
SDP

Catalog No. 19161600
Encompass
Encompass_Preferred
Change view
Click to view available options
Product Type:
Storage Cover
Material:
Vinyl
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type Material
19-161-600 Storage Cover Vinyl
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-161-600 Supplier Ferno™ Supplier No. 0314023
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Storage options and brackets for use with Ferno EZ Glide Evacuation Chair

For Evacuation Chair, see cat. no. 19161597.

Specifications

Product Type Storage Cover
Material Vinyl
For Use With (Equipment) EZ glide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.