Learn More
ABclonal Technology Ferredoxin Reductase
SDP
Supplier:  ABclonal Technology A27716100UL
Encompass_Preferred
This gene encodes a mitochondrial flavoprotein that initiates electron transport for cytochromes P450 receiving electrons from NADPH Multiple alternatively spliced transcript variants have been found for this gene
Catalog No. 50-000-12655