Learn More
Sino Biological Anti-CD4 Antibody, Mouse Monoclonal
Supplier:  Sino Biological 60003MM0820
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Ferret CD4 ( Catalog#60003-F08H; ABS50090.1; Met1-Leu401). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-240-8285