Learn More
Sino Biological Recombinant Anti-CD8 alpha Antibody (PerCP), Rabbit Monoclonal
Supplier:  Sino Biological 60001R082C100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Ferret CD8a / Lyt2 (rF CD8a / Lyt2; Catalog#60001-F08H; ABS50091.1; Met1-Glu186) and conjugated with PerCP under optimum conditions, the unreacted PerCP was removed.
Catalog No. 50-241-1512