Learn More
Kingfisher Biotech Inc Ferret IL-2 ELISA
SDP
Supplier:  Kingfisher Biotech Inc DIY1931FT004
Encompass_Preferred
The Ferret IL-2 ELISA contains capture antibody standard and detection antibody for development of a Ferret IL-2 ELISA. The antibodies have been determined to function in an ELISA with the standard provided. Optimal buffers concentrations incubation times incubation temperatures and methods for the ELISA have not been determined. This allows for complete flexibility to provide optimal dynamic range and sensitivity based on your sample type. Alternate names include IL2 IL-2 TCGF lymphokine interleukin 2. The quantities of components provided are not matched. Components may also be purchased separately. For research use only. Made in the USA
Catalog No. 50-228-5604