Learn More
Kingfisher Biotech Inc Human/Monkey VEGF-A ELISA - Less than or equal to 10 plates (96-well)
SDP
Supplier:  Kingfisher Biotech Inc DIY1956FT004
Encompass_Preferred
The Human/Monkey VEGF-A ELISA contains capture antibody, standard, and detection antibody for development of a Human/Monkey VEGF-A ELISA. The antibodies have been determined to function in an ELISA with the standard provided. Optimal buffers, concentrations, incubation times, incubation temperatures, and methods for the ELISA have not been determined. This allows for complete flexibility to provide optimal dynamic range and sensitivity based on your sample type. The Human/Monkey VEGF-A ELISA can also be used to measure Sooty Mangabey, Green Monkey, Peter’s Angolan Colobus, Western Lowland Gorilla, Rhesus Monkey, Pig-tailed Macaque, Drill, Northern White-cheeked Gibbon, Pygmy Chimpanzee, Chimpanzee, Olive Baboon, and Golden Snub-nosed Monkey VEGF-A. Alternate names include VEGFA, MVCD1, VEGF, VPF, vascular endothelial growth factor A. The quantities of components provided are not matched. Components may also be purchased separately. For research use only. Made in the USA
Catalog No. 50-228-5620