Learn More
Kingfisher Biotech Inc Ferret/Mink PDGF-BB Protein (Yeast, Endotoxin - Free) - 25 ug
SDP
Catalog No. 502964718
Encompass
The Ferret/Mink PDGF-BB yeast-derived recombinant protein is not tagged and is naturally endotoxin-free. The purity is greater than 98% as visualized by SDS-PAGE analysis. Ferret/Mink PDGF-BB applications are for cell culture, ELISA standard, and Western Blot Control. Ferret/Mink PDGF-BB yeast-derived recombinant protein can be purchased in multiple sizes. Ferret/Mink PDGF-BB Specifications: (Molecular Weight:18.3 kDa) (Amino Acid Sequence:SLGSPTVAEP AMIAECKTRT EVFEISRRLI DRTNANFLVW PPCVEVQRCS GCCNNRNVQC RPTQVQLRPV QVRKIEIVRK KPTFKKATVT LEDHLACKCE TVVATRPVTR SPGSTQEQRA AKTPQTRVTI RTVRVRRPPK GKHRKFKHTH DKKALKETLG A). For research use only. Made in the USA
Catalog No. 50-296-4718 Supplier Kingfisher Biotech Inc Supplier No. RP2227FT025
The Ferret/Mink PDGF-BB yeast-derived recombinant protein is not tagged and is naturally endotoxin-free. The purity is greater than 98% as visualized by SDS-PAGE analysis. Ferret/Mink PDGF-BB applications are for cell culture, ELISA standard, and Western Blot Control. Ferret/Mink PDGF-BB yeast-derived recombinant protein can be purchased in multiple sizes. Ferret/Mink PDGF-BB Specifications: (Molecular Weight:18.3 kDa) (Amino Acid Sequence:SLGSPTVAEP AMIAECKTRT EVFEISRRLI DRTNANFLVW PPCVEVQRCS GCCNNRNVQC RPTQVQLRPV QVRKIEIVRK KPTFKKATVT LEDHLACKCE TVVATRPVTR SPGSTQEQRA AKTPQTRVTI RTVRVRRPPK GKHRKFKHTH DKKALKETLG A). For research use only. Made in the USA