Learn More
Sino Biological Anti-CD20 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 60004T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Ferret CD20 (Catalog#60004-F08H; Glu213-Pro297). CD20 specific IgG was purified by Ferret CD20 affinity chromatography.
Catalog No. 50-255-2929